Would you like to log in again?
EnzyChrom™ Hexokinase Assay Kit
Supplier: BioAssay Systems
EnzyChrom™
EHXK-100
77441-684EA 459 USD InStock
77441-684
EnzyChrom™ Hexokinase Assay Kit
Assays Enzyme Assays
Hexokinase activity determination in biological samples (e.g. microorganisms, plasma, serum, cultured cells, tissue and culture media).
  • Fast and sensitive
  • Linear detection range (20 μl sample): 0.22 to 100 U/L for 15 min reaction
  • Convenient
  • Homogeneous 'mix-incubate-measure' type assay
  • High-throughput
  • Can be readily automated on HTS liquid handling systems for processing thousands of samples per day
HEXOKINASE (E.C. 2.7.1.1) is a mitochondrial and cytosolic enzyme and the first committed step in the glycolysis pathway. Hexose sugars, mainly glucose, are phosphorylated using ATP to form glucose-6-phosphate. Hexokinases are implicated in a myriad of human diseases, including cancer and diabetes. BioAssay Systems’ non-radioactive, colorimetric Hexokinase assay involves a stepwise reaction based on the reduction of the tetrazolium salt MTT in an NADPH-coupled enzymatic reaction to a colored product which exhibits an absorption maximum at 565 nm. The increase in absorbance at 565 nm is proportional to the enzyme activity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty