Would you like to log in again?
EnzyFluo™ Inosine Assay Kit
Supplier: BioAssay Systems
EnzyFluo™
EINS-100
77441-688EA 429 USD InStock
77441-688
EnzyFluo™ Inosine Assay Kit
Assays Enzyme Assays
For determination of inosine concentration in cell lysate, serum, and other biological samples.
  • Sensitive and accurate
  • Use as little as 20 μl of sample
  • Linear detection range in 96-well plate for 30 min incubation with 1 to 25 μM
  • Simple and convenient
  • The procedure involves addition of a single working reagent and incubation for 30 min at room temperature
  • Fast and high-throughput
  • Assays using 96-well plates and liquid handling system could allow simultaneous processing tens of thousands of samples per day
INOSINE is a purine nucleoside formed from Ribose and Hypoxanthine that can be found in most animal tissues and fluids. Inosine is most often found in tRNAs and is important for proper translation of the genetic code. It may play a role in cancer immunometabolism. It is degraded via the purine degradation pathway, first by purine nucleoside phosphorylase to hypoxanthine, then to xanthine by xanthine oxidase and then finally to uric acid by further action of xanthine oxidase.
Simple, direct and high-throughput assays for measuring inosine find wide applications in research and drug discovery. BioAssay Systems’ inosine assay kit uses a single working reagent that combines the enzyme reactions and color reaction in one step. The change in fluorescence intensity at λex/em = 530/585 nm is directly proportional to inosine concentration in the sample.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty