Would you like to log in again?
EnzyChrom™ Phosphoglucomutase Assay Kit
Supplier: BioAssay Systems
EnzyChrom™
EPGM-100
77441-690EA 539 USD InStock
77441-690
EnzyChrom™ Phosphoglucomutase Assay Kit
Assays Enzyme Assays
For quantitative determination of phosphoglucomutase enzyme activity in biological samples.
  • Safe and sensitive
  • Non-radioactive assay
  • Linear detection range in 96-well plate: 0.5 to 20 U/L activity
  • Fast and convenient
  • The procedure involves addition of a single working reagent and incubation for 20 min
  • Room temperature assay
  • No 37 °C incubator is needed
  • High-throughput
  • Homogeneous 'mix-incubate-measure' type assay
  • Can be readily automated to assay thousands of samples per day
Phosphoglucomutase (E.C. 5.4.2.2) is an enzyme which catalyzes the interconversion of glucose-1-phosphate (G1P) and glucose-6-phosate (G6P). Phosphoglucomutase (PGM) plays a crucial role in glycogen synthesis and degradation. PGM 1 deficiency can cause a rare congenital disorder of glycosylation. A decrease in PGM activity has also been linked with oxidative stress.
In BioAssay Systems EPGM-100 assay, PGM converts G1P to G6P, which is then oxidized by glucose-6-phosphate dehydrogenase. The formed NADPH is coupled to a formazan chromogen. The increase in absorbance at 460 nm is directly proportional to phosphoglucomutase activity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty