Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CORIO™ CP Refrigerated/Heating Circulators

Supplier: Julabo

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CORIO™
9013715.N1.33 9013714.S1.33 9013714.S1.02 9013704.02 9013708.33 9013706.02 9013728.N1.02 9013713.S1.33 9013728.N1.33 9013705.33 9013701.33 9013703.13 9013703.02 9013705.02 9013707.02 9013708.33 9013702.02 9013701.N1.02 9013707.02 9013705.33 9013727.02 9013702.33 9013703.33 9013714.N1.33 9013727.33 9013701.N1.33 9013701.02 9013702.33 9013707.33 9013717.N1.33 9013706.33 9013707.33 9013715.N1.02 9013704.33 9013704.02 9013705.02 9013713.S1.02 9013706.02 9013701.02 9013717.N1.02 9013713.N1.02 9013714.N1.02 9013702.02 9013706.33 9013701.33 9013713.N1.33 9013703.02 9013704.33
76558-220EA 7718.01 USD InStock
76558-220 77442-082 CA76353-848 76353-836 76353-858 76558-168 76353-838 CA76353-846 77442-080 76558-180 76558-160 76353-860 CA76353-854 76353-844 76558-236 77442-086 76353-846 CA76353-852 76558-238 77442-084 76353-840 76353-862 CA76353-850 76558-218 76353-864 76353-842 77442-088 CA76353-838 76558-198 CA76353-858 76353-848 CA76353-836 76558-254 CA76353-856 77442-090 76558-246 76353-854 CA76353-844 CA76353-864 CA76353-842 76353-856 77442-078 CA76353-862 76353-850 CA76353-840 77442-076 76353-852 CA76353-860
CORIO™ CP Refrigerated/Heating Circulators
Circulating Thermostats Open Circulator Systems Heating/Cooling Circulators
CORIO™ CP refrigerated/heating circulator combinations with stainless steel bath tanks for internal or external temperature applications from –50 to +200 °C.

  • Infinitely adjustable pressure pump
  • Easy to switch between internal and external circulation
  • Bright LED display
  • USB and RS232 ports

For internal and external temperature applications. High quality stainless steel bath tanks with bath lid and drain tap.

CORIO CP circulators feature a bright LED display button controls. A lever below the display enables simple adjustment between internal and external fluid flow. The USB and RS232 ports facilitate external communication to PC or PLC systems. Pump connections: M16×1 male threads.

To adapt to your individual application there is comprehensive range of accessories.

Certifications: CE, UKCA and DIN 12876-1 Class III.

Ordering information: Each unit is supplied with a pair of 8 and 12 mm hose barbs. Fluid, adapters, tubing sold separately.

Delivery information: Includes CORIO CP immersion circulator, refrigeration unit, mounting bridge, bath tank lid, pair of 8 and 12 mm hose barbs, integrated drain tap.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty