Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

MAGIO™ MS Refrigerated and Heating Circulators

Supplier: Julabo

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

MAGIO™
9032728.N1.33 9032727.33 9032707.33 9032728.N1.02 9032713.N1.02 9032715.N1.02 9032705.02 9032714.S1.02 9032714.N1.33 9032713.S1.33 9032715.N1.33 9032704.33 9032713.S1.02 9032714.N1.02 9032714.S1.33 9032705.33 9032704.02 9032713.N1.33 9032707.02 9032727.02 9032717.N1.02 9032706.02 9032717.N1.33 9032706.33
76558-194EA 11610.51 USD InStock
76558-194 76558-196 76558-188 76558-210 76558-222 76558-270 77442-148 76477-138 76477-136 77442-146 76477-148 76477-146 76477-154 76477-144 77442-140 76477-152 76477-142 76477-150 77442-150 76477-140 76558-228 76558-206 77442-144 77442-142
MAGIO™ MS Refrigerated and Heating Circulators
Circulating Thermostats Open Circulator Systems Heating/Cooling Circulators
The MAGIO™ series of cooling and heating circulators operate with a working temperature range of −50 to +300 °C and heating capacity of up to 3 kW. With their powerful pressure/suction pump the MAGIO™ units are ideal for external temperature applications. An extensive range of accessories and excellent dynamics mean that the MAGIO™ circulators can be modularly and individually adapted to applications.

  • Integrated programmer allows temperature time profiles to be run
  • Stainless steel wetted parts mean units are ideal for demanding external applications
  • Automatic shut off in the event of high temperature or low liquid level to avoid any potential damage
  • Bright, easy to read display
  • Bath tanks have high quality thermal insulation to save energy and a built in drain tap
  • Powerful, continuously adjustable pressure/suction pump

The high resolution, modern colour TFT touch display has a multi-lingual user interface and shows all important information at a glance. Three large, predefined main screens clearly display data and graphics with various application priorities. All display functions are easy to operate with a fingertip.

Modern interfaces make remote control easy and practical data management possible as well as allowing integration into process structures. All MAGIO™ units have integrated PT100 connection, USB interface, RS232 and Ethernet. Analogue interfaces are available as optional accessories.

Max. flow rate / max. pressure:
31 L/min, 0.92 bar (pressure), 0.4 bar (suction)

MS-310F is available with a natural refrigerant (article JULA9032743)

Certifications: CE, UKCA. DIN 12876-1 Class III.

Ordering information: Each unit is supplied with a pair of 8 and 12 mm hose barbs. Fluid, adapters, tubing sold separately.

Caution: Cooling capacity at +20; 0; −10; −20; −30 and −40 °C.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty