Would you like to log in again?
Microwave/Refrigerator Combinations with Allocator
Supplier: Felix Storch Inc
MRF29KA MRF708BLSSA
77442-216EA 1133 USD InStock
77442-216 77442-222
Microwave/Refrigerator Combinations with Allocator
Refrigerators
Microwave/Refrigerator combination with allocator with one-plug operation, ideal for hotels and other settings looking to maximize user convenience. The allocator will temporarily disable the power of the refrigerator while the microwave is in use, resuming after the cooking cycle is complete in order to protect against power surges.
  • USB ports
  • Reversible doors (refrigerator)
  • Stainless steel handle (microwave)
  • Mute function
  • Eco mode
  • Thin-line design
Environmentally friendly hydrocarbon-based refrigerant for a greener footprint that meets current CARB regulations.
Certifications: CARB compliant.
Packaging: Ships on one pallet in three boxes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty