Would you like to log in again?
Tryptic Soy Agar Plates, Teknova
Supplier: Teknova
T0144 T0144 T0144
100217-300PK 88.6 USD InStock
100217-300 CA100217-300 77442-968
Tryptic Soy Agar Plates, Teknova
Media for Microbiology Prepared Media for Microbiology
For isolating and cultivating a variety of fastidious microorganisms.
  • 20 Plates
  • Sterile
Contains 1.5% Tryptone, 0.5% soytone, 0.5% NaCl, and 1.5% agar.
Ambient shipping conditions. Store at room temperature.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty