Would you like to log in again?
R2A Agar Plates, Teknova
Supplier: Teknova
R1035 R1035 R1035
100216-352PK 136.99 USD InStock
100216-352 CA100216-352 77443-032
R2A Agar Plates, Teknova
Media for Microbiology Prepared Media for Microbiology
R2A Agar is used for the growth of heterotrophic organisms found in potable water.
  • 20 Plates
  • Sterile
R2A Agar Plates (per Liter): 0.05% Yeast Extract (0.5 g/L), 0.05% Proteose Peptone #3 (0.5 g/L), 0.05% Casamino Acids (0.5 g/L), 0.05% Dextrose (0.5 g/L), 0.05% Soluble Starch (0.5 g/L), 0.03% Sodium Pyruvate (0.3 g/L), 0.03% Dipotassium Phosphate (0.3 g/L), 0.005% Magnesium Sulfate (0.05 g/L), 1.5% Agar (15.0 g/L)
Ship and store at room temperature.
Final pH: 7.2 ± 0.2 (25°C).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty