Would you like to log in again?
Nutrient Agar Plates, Teknova
Supplier: Teknova
N0101 N0101 N0101
77443-106EA 126.85 USD InStock
77443-106 CA100219-190 100219-190
Nutrient Agar Plates, Teknova
Media for Microbiology Dehydrated Media for Microbiology
This nutrient agar is used as a general purpose medium for the examination of water and dairy products. Also used for the cultivation of fastidious microorganisms.
It contains 0.3% Beef extract, 0.5% peptone, 0.8% NaCl and 1.5% agar.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty