Would you like to log in again?
LB Agar Plates with 50 µg/ml Kanamycin, Teknova
Supplier: Teknova
L2025-4PK L5025 L1025 L3025 L1025 L1025 L5025 L2025 L3025
100218-230PK 217.1 USD InStock
100218-230 100216-588 CA100216-588 CA100218-230 77443-346 100218-444 CA100217-254 100217-254 CA100218-444
LB Agar Plates with 50 µg/ml Kanamycin, Teknova
Media for Microbiology Prepared Media for Microbiology
For growing strains of recombinant E. coli transformed with plasmids containing the resistance gene for Kanamycin (kan).
  • Sleeve of 20 plates
  • Sterile
Ingredients: 1.0% Tryptone (10.0 g/L), 0.5% Yeast Extract (5.0 g/L), 1.0% Sodium chloride (NaCl) (10.0 g/L), 1.5% Agar (15.0 g/L), 50 µg/mL Kanamycin.
Ambient shipping conditions. Store at 2 to 8 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty