Would you like to log in again?
LB Agar Plates with 25 µg/ml Kanamycin, Teknova
Supplier: Teknova
L5023 L1023 L5623 L1023 L1023 L3023 L5623 L5023 L3023
101414-080PK 163.72 USD InStock
101414-080 100216-584 CA100218-226 CA100216-584 CA100217-250 100217-250 100218-226 77443-350 CA101414-080
LB Agar Plates with 25 µg/ml Kanamycin, Teknova
Media for Microbiology Prepared Media for Microbiology
For use with kanamycin resistant strains and strains transformed with plasmids such as pPIC9K, TOPO-TA, TOPO pCR-Blunt, and pUC9P.
  • Sleeve of 20 plates
  • Sterile
Ingredients: 1.0% Tryptone (10.0 g/L), 0.5% Yeast Extract, (5.0 g/L), 1.0% Sodium Chloride (10.0 g/L), 1.5% Agar (15.0 g/L), 25 µg/ml Kanamycin.
Room temperature shipping conditions. Store at 2 to 8 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty