Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

LB Agar Plates with Chloramphenicol, Teknova

Supplier: Teknova
L1016 L1015 L1016 L1016 L1015
77443-356EA 82.38 USD InStock
77443-356 100216-572 100216-570 CA100216-572 CA100216-570
LB Agar Plates with Chloramphenicol, Teknova
Media for Microbiology Prepared Media for Microbiology
For use with chloramphenicol resistant strains and strains harboring plasmids such as pBC KS+/-, pJAK17, pMMB208 series plasmids.

  • Sleeve of 20 plates
  • Sterile

Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% agar chloramphenicol.

Ambient shipping conditions. Refrigerate to store.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty