Would you like to log in again?
LB Agar Plates with 50 µg/ml Carbenicillin, Teknova
Supplier: Teknova
L1008 L5008 L3008 L2008 L1008 L1008
77443-372EA 58.85 USD InStock
CA100216-558 CA100218-198 CA100217-222 CA100218-410 77443-372 100216-558
LB Agar Plates with 50 µg/ml Carbenicillin, Teknova
Media for Microbiology Prepared Media for Microbiology
Carbenicillin is an analog of ampicillin. For use with ampicillin resistant strains and strains harboring plasmids such as pBluescript, pGEM, pUC series plasmids.
  • Sleeve of 20 plates
  • Sterile
Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% agar 50 µg/ml carbenicillin.
Ambient shipping conditions. Refrigerate to store.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty