Would you like to log in again?
LB Agar Plates with 100 µg/ml Ampicillin, Teknova
Supplier: Teknova
L5004 L1004 L2004 L1004 L5004 L1004
77443-376EA 58.85 USD InStock
77443-376 CA100217-214 100217-214 CA100218-402 100216-550 CA100216-550
LB Agar Plates with 100 µg/ml Ampicillin, Teknova
Media for Microbiology Prepared Media for Microbiology
For use with recombinant strains of E.coli containing plasmids with the resistance gene to Ampicillin (bla or ampR) such as pBluescript, pGEM, or the pUC series of plasmids.
  • Sleeve of 20 plates
  • Sterile
Ingredients: 1.0% tryptone (10.0 g/l), 0.5% yeast extract (5.0 g/l), 1.0% sodium chloride (NaCl) (10.0 g/l), 1.5% agar (15.0 g/l), 100 µg/ml ampicillin.
Ambient shipping conditions. Store at 2 to 8 °C.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty