Would you like to log in again?
LB Agar Plates with 50 µg/mL Ampicillin, Teknova
Supplier: Teknova
L1002 L1002 L5002 L5002 L1002
77443-378EA 126.85 USD InStock
CA100216-546 77443-378 CA100217-210 100217-210 100216-546
LB Agar Plates with 50 µg/mL Ampicillin, Teknova
Media for Microbiology Prepared Media for Microbiology
For use with recombinant E. coli strains harboring plasmids such as pBluescript, pGEM, and the pUC series.
  • 20 plates
  • Sterile
Ingredients: 1.0% Tryptone (10.0 g/L), 0.5% Yeast Extract, (5.0 g/L), 1.0% Sodium Chloride (10.0 g/L), 1.5% Agar (15.0 g/L), 50 µg/ml Ampicillin.
Room temperature shipping conditions. Store at 2 to 8 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty