Would you like to log in again?
Brain-Heart Infusion Agar Plates, Teknova
Supplier: Teknova
B1010 B1010 B1010
100216-532PK 125.79 USD InStock
100216-532 CA100216-532 77443-496
Brain-Heart Infusion Agar Plates, Teknova
Media for Microbiology Prepared Media for Microbiology
Brain Heart Infusion (BHI) Agar is a general-purpose medium suitable for the cultivation of a wide variety of organisms, including bacteria, yeasts and molds.
  • Sleeve of 20 plates
  • Sterile
Contains 3.7% Brain Heart Infusion Powder (37.0 g/L), 1.5% Agar (15.0 g/L)
Ambient shipping conditions. Store at room temperature.
Final pH of 7.4.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty