Would you like to log in again?
TSA Plates with 5% Sheep Blood, Teknova
Supplier: Teknova
B0142 B0142 B0142
77443-500EA 183.08 USD InStock
77443-500 100220-036 CA100220-036
TSA Plates with 5% Sheep Blood, Teknova
Media for Microbiology Prepared Media for Microbiology
Used in microbiological environmental monitoring.
  • Sleeve of 20 plates
  • Sterile
Contains 1.5% Tryptone, 0.5% soytone, 0.5% sodium chloride, 1.5% agar, and 5% sheep's blood.
Ambient shipping conditions. Refrigerate to store.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty