Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EZBio® Titanium Round (TR) Polycarbonate Carboys

Supplier: Foxx Life Sciences
EZBio®
518-CH52-FLS 518-CH51-FLS 518-CG52-FLS 518-CG51-FLS 518-CF52-FLS 518-CF51-FLS
77447-854EA 433.56 USD InStock
77447-854 77107-004 77107-002 77107-008 77107-006 77447-852
EZBio® Titanium Round (TR) Polycarbonate Carboys
Bottles Carboys
Foxx Life Sciences’ EZBio® Titanium Round (TR) Polycarbonate Carboys are molded with ultra-pure USP Class VI resins, FDA 21 CFR 177 Grade materials. They are supplied double bagged, and are available sterile or non sterile.

  • Available in 5 and 10 L sizes
  • Made entirely of 100% USP Class VI components
  • Compatible with leakproof VersaCap® 83B caps for flexibility of interchangeable connectors
  • Carboys can withstand low temperatures down to −135 °C
  • Manufactured in a Class 7 cleanroom
  • Autoclavable

EZBio® Titanium Round (TR) Carboys are advanced fluid handling systems developed with the end user in mind. The EZBio® TR Carboy serves as a highly versatile container suitable for all fluid management applications. These round carboys are perfect for applications that require homogeneous mixing. The shape eliminates any pockets of undissolved material that square carboys might develop due to uneven corners.

Certifications: USP Class VI components. Manufactured in an ISO 13485 facility in North America.

Packaging: Bottles are packaged in shrinked-wrapped modules.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty