Would you like to log in again?
Diamond® Essentials™ Griffin Beakers
Supplier: Globe Scientific
Diamond® Essentials™
3650-250 3652-10M 3650-10M 3652-50 3652-2M 3650-100 3652-5M 3652-1M 3650-50 3652-100 3650-2M 3650-600 3652-600 3652-250 3650-1M 3650-5M
77447-890EA 28.5 USD InStock
77447-890 77447-880 77447-892 77447-882 77447-894 77447-884 77447-896 77447-886 77447-898 77447-888 77447-878 77447-900 77447-902 77447-904 77447-906 77447-908
Diamond® Essentials™ Griffin Beakers
Beakers
Diamond® Essentials™ griffin style beakers meet the demands for mixing, pouring, stirring, and measuring solutions in many laboratory operations.
  • Manufactured from virgin, USP class VI resins
  • Bold, printed, dual scale graduations
  • Single non-drip spout
  • Autoclavable at 121 °C
  • Chemically resistant to most acids, bases, and many common solvents
These beakers are the safe, shatterproof alternative to glass in operations where plastic is suitable. Value, quality, and performance are insured by precision molding in an automated manufacturing process. The beakers are designed with a single, no drip spout and the beakers with handles facilitate easy pouring.
Available in economic polypropylene (PP) or crystal clear polymethylpentene (PMP).
Certifications: Complies with FDA 21-CFR and European Pharmacopeia.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty