Would you like to log in again?
Diamond® Essentials™ Griffin Beakers, with Handle
Supplier: Globe Scientific
Diamond® Essentials™
3654-2M 3656-600 3656-5M 3656-2M 3654-5M 3654-600 3654-1M 3656-100 3656-250 3654-100 3654-250 3656-1M
77447-930EA 85.03 USD InStock
77447-930 77447-920 77447-932 77447-910 77447-922 77447-912 77447-924 77447-914 77447-926 77447-916 77447-928 77447-918
Diamond® Essentials™ Griffin Beakers, with Handle
Beakers
These Griffin style beakers meet the demands for mixing, pouring, stirring, and measuring solutions in many laboratory operations.
  • Manufactured from virgin, USP Class VI resins
  • Bold, printed, dual scale graduations
  • Single non-drip spout
  • Autoclavable at 121 °C
  • Chemically resistant to most acids, bases, and many common solvents
  • Beakers without handles stack to save storage space
These beakers are the safe, shatterproof alternative to glass in operations where plastic is suitable. Value, quality, and performance are insured by precision molding in an automated manufacturing process. These beakers are designed with handles that facilitate easy pouring.
Available in economic polypropylene (PP) or crystal clear polymethylpentene (PMP).
Certifications: Complies with FDA 21-CFR and European Pharmacopeia.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty