Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Tacky Traxx™ PVC Cleanroom Sticky Mat Frames
Supplier: High-Tech Conversions
Tacky Traxx™
TTF-0-3761 TTF-0-3737 TTF-0-2746 TTF-0-1937 TTF-0-2537
77448-026EA 254.34 USD InStock
77448-026 77448-024 77448-022 77448-020 77448-018
Tacky Traxx™ PVC Cleanroom Sticky Mat Frames
Floor Mats Mat Frames
Tacky Traxx™ PVC cleanroom sticky mat frames are an indispensable tool for contamination control. Made from durable PVC, these economical frames provide a designated area for your cleanroom sticky mats, allowing you to effortlessly clean underneath and move them to various sections of your facility as needed.
  • Lightweight frame allows for easy cleaning underneath
  • Provides designated areas for tacky mats
  • Economical mat frame
  • Lightweight
  • Made of durable PVC
  • Non-skid mesh backing
Crafted from lightweight yet robust PVC, the Tacky Traxx™ PVC Cleanroom Sticky Mat Frame can withstand the rigors of daily use in demanding cleanroom environments. Its non-skid mesh backing provides unwavering stability, preventing unwanted movement and ensuring it remains firmly in place, even in high-traffic areas.
Enables full-floor surface disinfecting. With a simple lift of the frame, you gain access to the floor beneath, allowing for thorough surface disinfecting. This meticulous attention to detail ensures that your cleanroom entry areas remain immaculate, meeting the most stringent standards.
Packaging: Sold individually.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty