Would you like to log in again?
PG-Seq™ Rapid Kit V2 and Index Primers
Supplier: Revvity Health Sciences
PG-Seq™
4341-IL48A 4340-0248 4341-IL48B 4341-IL48B 4361-IL96B 4361-IL96D 4361-IL96C 4340-0248 4361-IL96A 4361-IL96B 4361-IL96D 4361-IL48A 4361-IL48B 4361-IL96A 4341-IL48A 4361-IL96C 4361-IL48A 4361-IL48B
77448-078EA 1625 USD InStock
CA77448-078 77448-078 CA77448-076 CA77448-086 77448-076 CA77448-074 77448-074 CA77448-084 77448-086 CA77448-072 77448-072 CA77448-082 77448-084 CA77448-070 77448-070 CA77448-080 77448-082 77448-080
PG-Seq™ Rapid Kit V2 and Index Primers
Nucleic Acid Reagents Next Generation Sequencing Reagents
The PG-Seq™ Rapid kit V2 has been developed to analyze picogram quantities of DNA (single/multi-cells or low template DNA) from an embryo biopsy for preimplantation genetic testing.
  • Updated, streamlined, simple PGT protocol
  • Accurate copy number detection
  • Flexible kit format
The PG-Seq™ Rapid kit V2 has been developed to analyze picogram quantities of DNA (single/multi-cells or low template DNA) from an embryo biopsy for preimplantation genetic testing. The PG-Seq™ Rapid kit v2 utilizes whole genome amplification (WGA) and next generation sequencing (NGS) technology to accurately screen all 24 chromosomes for whole chromosome aneuploidy and sub-chromosomal abnormalities.
Certifications: Manufactured in ISO 9001 facility.
Ordering information: PG-seq™ Index primers must be ordered with PG-seq kits.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty