Would you like to log in again?
NEXTFLEX™ 16S V4 Amplicon-Sequence 2.0 Kits
Supplier: Revvity Health Sciences
NEXTFLEX™
NOVA-4203-06 NOVA-4203-02 NOVA-4203-06 NOVA-4203-04 NOVA-4203-05 NOVA-4203-07 NOVA-4203-07 NOVA-4203-02 NOVA-4203-04 NOVA-4203-03 NOVA-4203-05 NOVA-4203-03
77448-102EA 2082 USD InStock
77448-102 CA77448-100 77448-100 CA77448-110 77448-110 CA77448-108 77448-108 CA77448-106 77448-106 CA77448-104 77448-104 CA77448-102
NEXTFLEX™ 16S V4 Amplicon-Sequence 2.0 Kits
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEXTFLEX® 16S V4 Amplicon-Seq Kit 2.0 is designed to prepare multiplexed amplicon libraries that span the fourth hypervariable domain of microbial 16S ribosomal RNA (rRNA) genes.
  • Fast and easy library prep protocol
  • Low input – Only 1 ng of input required
  • Multiplex up to 384 libraries
  • Custom sequencing primers are not required
  • Automation protocols are now available for the Sciclone® NGS and NGSx Workstations to automate your 16S sequencing
  • Functionally tested with Illumina® sequencing platforms
Optimized Protocol Offers Lower PCR Bias and Fewer Off-target Reads
The NEXTFLEX 16S V4 Amplicon-Seq Kit 2.0 incorporates a second PCR step in the protocol for the addition of the sample-specific index which reduces off-target reads encountered during amplicon sequencing.
384 Barcodes Available for Cost-effective 16S V4 rRNA Sequencing
The NEXTFLEX 16S V4 Amplicon-Seq Library Prep Kit 2.0 can be used for multiplexing up to 384 samples, to greatly reduce the cost of sequencing. The barcodes are offered in sets of 4, 12, 48 or 96 barcodes and supplied with 2 reactions worth of each barcode.
Automate your 16S V4 Library Prep to Increase Throughput and Reduce Errors
A NEXTFLEX® automation protocol is now available on the Sciclone® NGS and NGSx workstations to help labs increase their throughput and reduce human errors.
Certifications: Manufactured in ISO 9001 facility.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty