Would you like to log in again?
NEXTFLEX™ RNA-Sequence 2.0 UDI Barcodes
Supplier: Revvity Health Sciences
NEXTFLEX™
NOVA-512922 NOVA-512920-EVAL16 NOVA-512920-EVAL48 NOVA-512922 NOVA-512920 NOVA-512920 NOVA-512923 NOVA-512921 NOVA-512923 NOVA-512920-EVAL16 NOVA-512920-EVAL48 NOVA-512921
77448-124EA 1019 USD InStock
77448-124 CA77448-122 77448-122 CA77448-120 77448-120 CA77448-118 CA77448-116 CA77448-126 77448-118 77448-126 CA77448-124 77448-116
NEXTFLEX™ RNA-Sequence 2.0 UDI Barcodes
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEXTFLEX™ RNA-sequence 2.0 UDI barcodes are designed to prepare multiplexed single and paired-end cDNA libraries from total RNA, mRNA-enriched RNA or rRNA-depleted RNA for Illumina platforms.
  • UDI barcodes for multiplexing RNA-sequence libraries
  • Compatible with both paired-end and single-read sequencing
  • Every lot is tested for index purity and functionality by sequencing
The NEXTFLEX™ unique dual index barcodes contain two unique indexes on each adapter, with a hamming distance of at least three throughout the entire series, to allow error correction. The NEXTFLEX™ unique dual index RNA-sequence barcodes can be used for PCR-free RNA sequencing applications. Patterned flow cells found on the Illumina® Novasequence®, Hisequence® 3000/4000, and Hisequence® X platforms suffer from increased rates of sample mis-assignment during sequencing. The use of adapters with unique dual indices during sequencing prevents such mis-assigned reads from appearing in final data sets, allowing for the highest assurance of data integrity. Every lot is tested for index purity and functionality by sequencing.
Certifications: Manufactured in ISO 9001 facility.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty