Would you like to log in again?
NEXTFLEX™ Rapid DNA-seq 2.0 Kits
Supplier: Revvity Health Sciences
NEXTFLEX™
NOVA-5188-01 NOVA-5188-03 NOVA-5188-02 NOVA-5188-02 NOVA-5188-01 NOVA-5188-03
77448-194EA 2927 USD InStock
CA77448-194 CA77448-192 77448-194 CA77448-190 77448-192 77448-190
NEXTFLEX™ Rapid DNA-seq 2.0 Kits
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEXTFLEX® Rapid DNA-seq 2.0 kit is designed to prepare multiplexed libraries for single or paired-end sequencing using Illumina® platforms from fragmented DNA.
  • Complete library prep solution, including size selection beads
  • Up to 1536 UDI barcodes are available separately
  • Convenient and cost-effective package in an 8, 48, or 96 reaction format
  • Simplified workflow
  • Robust genome coverage
  • Reliable performance with GC-bias and sequencing bias mitigation
  • Low input requirement for genomic DNA down to 1 ng
  • Functionally tested with Illumina® sequencing platform
  • Automation compatible with liquid handlers, including Revvity's Sciclone® G3 NGSx workstations, Zephyr® G3 NGS workstation, and BioQule NGS system
The NEXTFLEX  Rapid DNA-seq kit 2.0 produces library preps with a larger number of unique sequencing reads, allowing for robust genome coverage in challenging GC-rich sequences that can introduce bias.
Certifications: Manufactured in ISO 9001 Facility.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty