Would you like to log in again?
NEXTFLEX™ Rapid Directional RNA-Sequence 2.0 Kits
Supplier: Revvity Health Sciences
NEXTFLEX™
NOVA-5198-02 NOVA-5198-03 NOVA-5198-53 NOVA-5198-01 NOVA-5198-43 NOVA-5198-43 NOVA-5198-01 NOVA-5198-03 NOVA-5198-02 NOVA-5198-53
77448-202EA 2162 USD InStock
CA77448-200 77448-202 CA77448-198 77448-200 CA77448-196 77448-198 77448-196 CA77448-204 CA77448-202 77448-204
NEXTFLEX™ Rapid Directional RNA-Sequence 2.0 Kits
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEXTFLEX® Rapid directional RNA-Sequence kit 2.0 is designed to prepare directional, strand specific RNA libraries for sequencing using Illumina® sequencers.
  • High coverage uniformity with low duplication rate
  • Optimized for use with 5 ng to 5 µg total RNA with reverse transcriptase and cleanup/size selection beads included
  • Simple protocol validated with NEXTFLEX® poly(A) beads 2.0 and NEXTFLEX® RiboNaut™ rRNA depletion kit (human, mouse, rat)
  • Designed to work with NEXTFLEX® RNA-Seq 2.0 Unique Dual Index barcodes that allow a wide range of multiplexing (2 up to 1,526 samples in one run)
  • Automated on the Sciclone® G3 NGSx and Zephyr® G3 NGS workstations
The NEXTFLEX® Rapid directional RNA-seq kit 2.0 includes reverse transcriptase, necessary library preparation reagents, and cleanup/size selection beads optimized to ensure reliable performance. The kit involves a simple library preparation protocol that has been tested with the NEXTFLEX® poly(A) beads 2.0 and NEXTFLEX® RiboNaut™ rRNA depletion kit (human, mouse, rat) to accommodate total RNA as input. The NEXTFLEX® rapid directional RNA-seq kit 2.0 is designed to be used with NEXTFLEX® RNA-Seq 2.0 Unique dual index barcodes (6.25 μm), which are color-balanced and have undergone proprietary purity QC metrics to generate reliable sequencing results for every sample.
Certifications: Manufactured in ISO 9001 facility.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty