Would you like to log in again?
NEXTFLEX™ Human Core Exome Kits
Supplier: Revvity Health Sciences
NEXTFLEX™
NOVA-6011-EVAL NOVA-6011-48 NOVA-6011-EVAL NOVA-6011-96 NOVA-6011-48 NOVA-6011-96
77448-224EA 8057 USD InStock
CA77448-222 77448-224 77448-222 CA77448-226 CA77448-224 77448-226
NEXTFLEX™ Human Core Exome Kits
Nucleic Acid Reagents Next Generation Sequencing Reagents
NEXTFLEX® Human core exome V2 (CES) panel is designed to overcome the limitations of analyzing genetic variants of interest with respect to conventional whole-exome sequencing.
  • Complete whole exome coverage
  • Exceptional target capture performance
  • Rapid same-day workflow
  • Automated on Sciclone® G3 NGSx workstation
  • Bead-based normalization
The NEXTFLEX® human CES panel includes significant genes used in other panels and additional genes, therefore allowing customers to obtain comprehensive information of mutations in genes of interest in a cost-effective manner. The 7513 genes covered in this panel can be reviewed in the NEXTFLEX CES panel gene list.
Certifications: Manufactured in ISO 9001 Facility.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty