Would you like to log in again?
CleanMax® Cleanroom Frocks
Supplier: Lakeland Industries
CleanMax®
CTL191CS-SM CTL191CM-SM CTL191CMB-SM
77449-034CS 425.03 USD InStock
77449-034 77449-030 77449-032
CleanMax® Cleanroom Frocks
Disposable Garments
Cleanroom garments are constructed and packaged to offer premium protection for your cleanroom and controlled environments.
  • Available in sterile and non-sterile configurations
  • Frock with zipper closure and collar
  • Bound seam
CTL191CM (clean manufactured) and CTL191CS (clean manufactured and sterile) cleanroom frock is ideal for ISO Class 4 to 6 cleanrooms.
Delivery information: These products are offered in a case of 30 with sizes small to 5XL available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty