Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Delta Carbin Program for PPE Recycling
Supplier: Polycarbin
PR01-04-01
77449-050PK 460.82 USD InStock
77449-050
Delta Carbin Program for PPE Recycling
Boxes Waste Disposal Boxes
Tired of unecessary waste in the lab? So are we. Polycarbin's Delta carbin program is an all-in-one mailback service for laboratory personal protective equipment (PPE).
  • Accepts all brands of PPE
  • Material stream is used to manufacture low-carbon lab products
  • Prepaid return labels and carbon neutral shipping
  • Access to personalized dashboard showing impact data
Polycarbin's Delta carbin program is a mailback service for PPE made from woven polypropylene (hair nets, gowns, booties, sleeves, etc.).
Simply place the 5-pack of carbins throughout the lab and collect eligible PPE. Once full, seal the carbin shut and set it out for UPS pick-up (prepaid return labels attached). Once recieved, your data will be uploaded to your dashboard so that you can measure your impact. The woven polypropylene stream will be sorted and processed, ultimately making its way back into our low-carbon lab products.
PPE containing metal clips or zippers, cloth, or paper are not accepted. PPE that comes into contact with the following is not eligible: Hazardous waste, pathological or surgical waste, infectious waste, viral waste (Exception, bacteriophage), spore-forming bacteria waste, human tissue waste animal tissue waste (Exception, hybridomas), fecal, urine, or saliva (Exception, Post-extraction/Inactivation).
Delivery information: Carbins come in packs of five. All carbins come with prepaid return labels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty