Would you like to log in again?
Polycarbin® LTS® Compatible Refill Towers
Supplier: Polycarbin
PC01-06-06R PC01-05-06R PC01-08-05 PC01-04-06R
77461-178CS 362.14 USD InStock
77461-178 77461-176 77461-182 77461-180
Polycarbin® LTS® Compatible Refill Towers
Pipette Tips
Polycarbin's LTS® compatible refill towers deliver the precision and accuracy you expect, with a fraction of the carbon footprint.
  • Wafer and rack made from 100% circular economy, post-consumer recycled content
  • Smaller carbon footprint compared to other refill and rack systems
  • Easy to transfer refills into polycarbin racks
  • Certified DNase, RNase and pyrogen free
  • Non-sterile (autoclavable)
Polycarbin's LTS® compatible refill system is designed to deliver an exceptional product that is less carbon and plastic-intensive. Refill tower housing is manufactured using post-consumer recycled content, and are shrink wrapped and packaged in an easy to recycle cardboard sleeve. Pipette tips do not contain recycled content and are made from virgin PP resin.
Certifications: Certified sterile and DNase, RNase and pyrogen free. Manufactured in an ISO 9001 compliant facility.
Ordering information: Note that Polycarbin's LTS® compatible refill towers do not come with racks (bases). These can be ordered separately by the sleeve. Please see part numbers 76635-484, 76635-488 and 76635-492.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty