Would you like to log in again?
BioXp® Select mRNA Synthesis Kits
Supplier: Telesis Bio
BioXP®
BX9-MRNA-RSU-CC-08 BX9-MRNA-RSM1-CC-48 BX9-MRNA-RSM1-CC-24 BX9-MRNA-RSM1-CC-08 BX9-MRNA-RSU-CC-48 BX9-MRNA-RSU-CC-24
77461-234EA 1440 USD InStock
77461-234 77461-236 77461-230 77461-232 77506-680 77461-228
BioXp® Select mRNA Synthesis Kits
Nucleic Acid Reagents RNA Synthesis and Transcription
The BioXp Select mRNA synthesis kit enables the synthesis of mRNA from an input DNA template.
  • mRNA size: 0.4 to 10 kb
  • Yield: 50 µg/well
  • Compatible with BioXp® 9600
  • Cap: 5' CleanCap® AG (3'OMe)
  • Modification: Pseudouridine
The BioXp® Select mRNA synthesis kit enables you to synthesize purified capped and tailed mRNA between 0.4 to 10 kb. The kit contains all the necessary reagents for on-demand synthesis of mRNA using a linear DNA template generated with a plasmid or PCR product for transcription.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty