Would you like to log in again?
BIOXP® 9600 Select Clone and AMP
Supplier: Telesis Bio
BIOXP®
BX9-SEL-CLNAMP-96 BX9-SEL-CLNAMP-24 BX9-SEL-CLNAMP-48 BX9-SEL-CLNAMP-16 BX9-SEL-CLNAMP-08
77461-256EA 2400 USD InStock
77461-256 77461-258 77461-262 77461-254 77461-260
BIOXP® 9600 Select Clone and AMP
Nucleic Acid Reagents Cloning Kits and Ligation
The Select DNA cloning and amplification kits generate a minimum of 10 µg of amplified DNA from fragments of 0.3 to 1.8 kb using the BioXp 9600.
  • Accepted input: Telesis Bio DNA fragments, IDT gBlocks™, Twist Biosciences fragments
  • Supports user-provided linearized vector
  • Length: 0.3 to 1.8 kb
  • Yield: >10 µg/well
  • Compatible with BioXp 9600
The BioXp Select DNA cloning and amplification kit contains Gibson Assembly® and other reagents necessary to synthesize at least 10 µg of amplified DNA from fragments and linearized vectors supplied by the user.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty