Would you like to log in again?
BIOXP® 9600 Select Plasmid AMP
Supplier: Telesis Bio
BIOXP®
BX9-SEL-PLSAMP-08 BX9-SEL-PLSAMP-96 BX9-SEL-PLSAMP-24 BX9-SEL-PLSAMP-48 BX9-SEL-PLSAMP-16
77461-266EA 1600 USD InStock
77461-266 77461-268 77461-264 77461-270 77461-272
BIOXP® 9600 Select Plasmid AMP
Nucleic Acid Reagents Cloning Kits and Ligation
BioXp Select plasmid amplification kits contain reagents for isothermal amplification of plasmid DNA.
  • Input: Plasmids ranging from 1.8 to 37 kb
  • Yield: >10 µg/well
  • Output format: Branched concatemers
  • Compatible with BioXp 9600
Contains reagents necessary to generate at least 10 micrograms of amplified DNA starting from a sequence verified plasmid supplied by the end user.
Caution: Branched concatemers need to be linearized prior to transfection. A recommended transfection protocol is included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty