Would you like to log in again?
BIOXP® Select mRNA
Supplier: Telesis Bio
BIOXP®
BX-MRNA-RSU-CC-08 BX-MRNA-RSM1-CC-24 BX-MRNA-RSM1-CC-08 BX-MRNA-RSM1-CC-48 BX-MRNA-RSU-CC-48 BX-MRNA-RSU-CC-24
77461-278EA 8640 USD InStock
77461-278 77461-274 77461-284 77461-276 77461-280 77461-282
BIOXP® Select mRNA
Nucleic Acid Reagents RNA Synthesis and Transcription
The BioXp Select mRNA synthesis kit enables the synthesis of mRNA from an input DNA template.
  • mRNA Length: 0.4 to 10 kb
  • Yield: >50 µg/well
  • Compatible with BioXp 3250
  • Cap: CleanCap® AG (3'OMe)
  • Modification: Pseudouridine
Contains reagents necessary to generate at least 10 micrograms of amplified DNA starting from a sequence verified plasmid supplied by the end user.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty