Would you like to log in again?
VWR® Sticky Mat
Supplier: Avantor
VWR®
30936259
77461-300EA 51.2 USD InStock
77461-300
VWR® Sticky Mat
Shakers and Mixers Shaking Incubators
Easily secure and remove samples without clamps.
  • Recommemded maximum speed not to exceed 250 rpm
  • Application temperature not to exceed 65 °C
  • Clean with water and air dry to remove adhesive quality
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty