Would you like to log in again?
VWR® Traceable® 7600 Smart Wi-Fi Data Logger
Supplier: Avantor
VWR®
07502-81
77463-148EA 294.65 USD InStock
77463-148
VWR® Traceable® 7600 Smart Wi-Fi Data Logger
Data Loggers
Stay connected to your critical environment measurements no matter where you are — get email, text, and push notifications to your internet-connected device.
  • Supports up to two smart probes and two door sensors
  • Smart Probes measure laboratory conditions such as temperature, and have built-in NIST calibration for quick set-up and easy replacement
  • Intuitive touchscreen display|simple, one-time setup only takes a few minutes
  • Enhance security with on-board measures including optional PIN protection to prevent tampering
  • Works with TraceableLIVE® cloud service for alerts, reporting, and configuration
These data loggers with Wi-Fi offer exciting features such as an intuitive touchscreen interface, enhanced PIN-based security, simple calibration with smart probes, door sensors to prevent accidental temperature excursions, faster setup, longer-lasting battery backup during power outages, support for 5 or 2.4 GHz Wi-Fi connections, and a cradle for mounting in place. Choose up to two smart probes and two door sensors for each logger. smart probes include temperature bullet probes, bottle probes, or vaccine bottle probes to meet a variety of applications.
Once setup is complete a user can securely monitor parameters 24/7, control alarms remotely, view data logging history, run reports in real time, and give access to other team members. No local software is required to use the cloud-based data interface. Get mobile push notifications, emails, or texts for out-of-range conditions, connectivity interruptions, low-battery condition, or calibration notice.
The logger can be mounted with the included hook-and-loop adhesive strip or placed on a countertop with the optional cradle. The cable on the smart probes allows a door gasket to close on them for use in refrigerators or freezers.
No. of channels: 4.
System requirements: 5 or 2.4 GHz Wi-Fi connection.
Note: A TraceableLIVE® subscription is required for use.
Certifications: NIST-traceable certificate of calibration from our A2LA-accredited ISO/IEC 17025:2017 (1750.01) laboratory.
Ordering information: A TraceableLIVE® subscription is required for use.
Delivery information: Mountable cradle, hook-and-loop adhesive strip, and power supply.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty