Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HyFlex® 11-812 Ultra-thin Light-duty Gloves

Supplier: Ansell
HyFlex®
837636
77463-298CS 427.02 USD InStock
77463-298
HyFlex® 11-812 Ultra-thin Light-duty Gloves
Gloves
HYFLEX® 11-812 ultra-thin light-duty gloves designed to pre-emptively tear if entangled in rotating tools.

  • Unique knit design: Gloves designed to tear when dangerously entangled
  • ZONZ™ Comfort Fit: Less stress and tension, more comfort and dexterity
  • Ultra-thin liner: Added tactile sensitivity, for precision handling
  • Liner color: White
  • Coating color: White
  • Coating material: Foam Nitrile
  • Construction: Knitted
  • Silicone free

Specialized protection: Designed for the operation of rotating machinery, HyFlex® 11-812 industrial gloves’ ultra-thin liner deliberately tears when they become entangled and the wearer is at risk of severe injury.

Advanced comfort and dexterity: Developed with ERGOFORM™ Technology and ZONZ™ Comfort Fit knitwork, these HyFlex® gloves offer users a tailored fit, for less strain and tension and greater comfort and dexterity.

Heightened tactility: Their thin liner also enhances tactile sensitivity, making them ideal for delicate handling.

These gloves can be used in primary industries like machinery and equipment, metal fabrication, automotive, automotive aftermarket, chemical, utilities, warehousing. Recommended for final assembly.

Certifications: They are Dermatest®-approved, solvent-free, and Oekotex®-certified for no harmful substances.

Caution: This glove will not directly protect the worker from injuries caused by contact with a sharp or high-speed rotating machine.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty