Would you like to log in again?
NIGHTSEA® Tru-Block Eyeshields
Supplier: Electron Microscopy Sciences
NIGHTSEA®
SFA-EYE-C SFA-EYE-S
77463-310EA 45.87 USD InStock
77463-310 77463-312
NIGHTSEA® Tru-Block Eyeshields
Microscopes
These soft, molded rubber high-sided microscope eye shields extend far enough to truly shield the eyes from any level of ambient light and eliminate distractions for improved viewing of fluorescence subject better.
  • Standard or Compact sizes are available in pairs
  • Standard size fits 36 to 45 mm (1.45 to 1.75")
  • Compact size fits 28 to 37 mm (1.10 to 1.46")
Light entering the eyes from the side can interfere with microscopy viewing in general, and fluorescence microscopy in particular. Eye cups are available, but standard cups don't extend far from the microscope and don't do a good job as ambient light increases. These soft, molded rubber high-sided microscope eye shields are the answer. The tall wings extend up far enough to truly shield the eyes from any level of ambient light and eliminate distractions for improved subject viewing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty