Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
NIGHTSEA® Stereo Microscope Fluorescence Adapters (SFA), Full System
Supplier: Electron Microscopy Sciences
NIGHTSEA®
SFA-RB-GO-DIM SFA-UV-DIM SFA-VI-DIM SFA-RB-DIM SFA-UV SFA-RB-PULSE SFA-GR-PULSE SFA-GR-DIM SFA-RB-GO-PULSE SFA-CY-PULSE SFA-CY-DIM SFA-UV-PULSE SFA-VI SFA-GR SFA-VI-PULSE
77463-340EA 3017.6 USD InStock
77463-340 77463-332 77463-344 77463-330 77463-342 77463-314 MSPP-SFARBGODI 77463-348 77463-346 102773-518 77463-308 77463-338 77463-316 77463-328 77463-306
NIGHTSEA® Stereo Microscope Fluorescence Adapters (SFA), Full System
Lights Fluorescent Lights, Lamps and Bulbs
The NIGHTSEA® stereo microscope fluorescence adapter system adapts almost all stereo microscopes for fluorescence viewing without needing modifications.
  • Lamp base with power supply and international plug set
  • Light head: ultraviolet, violet, royal blue, cyan, or green
  • Barrier filter
  • Viewing shield
  • Padded travel case
The modular design easily switches between different excitation/emission combinations to work with a variety of fluorescent proteins and other fluorophores.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty