Would you like to log in again?
Brady® ToughStripe® Max Floor Marking Tapes
Supplier: Brady
Brady®
177058 177078 177068 177015 177092 177076 177057 177024 177055 177036 177063 177083 177022 177062 177034 177035 177017 177069 177043 177016 177053 177023 177014 177060 177046 177074 177037 177026 177059 177045 177038 177056 177041 177096 177025 177052 177021 177070 177088 177019 177030 177042 177040 177054 177050 177032 177048 177047 177051 177020 177031 177049 177029 177028 177033 177027 177018
77463-482RL 175.77 USD InStock
77463-482 77463-480 77463-486 77463-442 77463-484 77463-440 77463-446 77463-402 77463-444 77463-488 77463-406 77463-404 77463-448 77463-508 77463-472 77463-470 77463-476 77463-432 77463-474 77463-430 77463-512 77463-436 77463-510 77463-434 77463-478 77463-514 77463-438 77463-460 77463-420 77463-464 77463-462 77463-424 77463-468 77463-502 77463-422 77463-466 77463-500 77463-428 77463-506 77463-426 77463-504 77463-408 77463-490 77463-494 77463-450 77463-492 77463-498 77463-454 77463-410 77463-496 77463-452 77463-458 77463-414 77463-412 77463-456 77463-418 77463-416
Brady® ToughStripe® Max Floor Marking Tapes
Tapes
Built stronger to last longer on punishing industrial floors. ToughStripe® Max floor marking tape is purpose-built to hold its ground in forklift and heavy traffic areas.
  • Permanent rubber based adhesive
  • Designed for indoor use
  • Gloss finish
  • Thickness: 0.024"
50% thinner than previous version of ToughStripe® Max and twice as tough as similarly thick vinyl tapes. Thinner profile and true edge-to-edge adhesive minimize pick points. Plasticizer-free polyester (PETG) material is more rigid than vinyl. Every layer is hand selected, pressure tested, and purpose built to protect colors and outlast the competition. Damage-isolating design keeps tape intact longer by preventing rips, gouges, and tears from running. Tape provides excellent color contrast for clear visibility under any lighting. Compact construction results in a smaller, lightweight roll. Can be easily installed by one person with minimal tools and is compatible with the ToughStripe® floor tape applicator.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty