Built stronger to last longer on punishing industrial floors. ToughStripe® Max floor marking tape is purpose-built to hold its ground in forklift and heavy traffic areas.
- Permanent rubber based adhesive
- Designed for indoor use
- Gloss finish
- Thickness: 0.024"
50% thinner than previous version of ToughStripe® Max and twice as tough as similarly thick vinyl tapes. Thinner profile and true edge-to-edge adhesive minimize pick points. Plasticizer-free polyester (PETG) material is more rigid than vinyl. Every layer is hand selected, pressure tested, and purpose built to protect colors and outlast the competition. Damage-isolating design keeps tape intact longer by preventing rips, gouges, and tears from running. Tape provides excellent color contrast for clear visibility under any lighting. Compact construction results in a smaller, lightweight roll. Can be easily installed by one person with minimal tools and is compatible with the ToughStripe® floor tape applicator.