Would you like to log in again?
Pig® Absorbent Sheet for Packaging with Liquid-Proof Backing
Supplier: New Pig
Pig®
PKG006
77463-546BG 100.45 USD InStock
77463-546
Pig® Absorbent Sheet for Packaging with Liquid-Proof Backing
Mailers and Shippers
Safely shipping liquids without damage or extra cleanup is easy using this absorbent sheet with leak-proof backing that prevents soaking through and damage to carton.
  • Liquid-proof backing prevents leaks from breaking through to outer packaging
  • Sheet absorbs free liquids and adds light cushioning
  • Backing's polyethylene construction improves puncture resistance
  • Handles most free liquids, including oils, coolants, solvents and water-based liquids
  • Soft, non-abrasive material won't scratch packed items
  • Sheet proven to hold together even when saturated
  • Sheets that are still dry upon shipment arrival are reusable for sustainability
Absorbency up to 11.4 gal. per bag, absorbency per pad up to 29.18 oz.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty