Would you like to log in again?
PIG® Aggressive Chemical Absorbent Packaging Bag
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PIG®
PKG3018 PKG3017 PKG3020 PKG3021 PKG3019
77463-570BX 231.36 USD InStock
77463-570 77463-564 77463-562 77463-568 77463-566
PIG® Aggressive Chemical Absorbent Packaging Bag
Bags Storage Bags
Secure your shipment from escaping aggressive chemicals by sealing your container in this packaging bag with absorbent sheet.
  • Sealable plastic bag with absorbent sheet traps and soaks up liquid escaping from container
  • Soft-but-heavyweight sheet adds light cushioning and scratch protection
  • Built-in mat flap lets you fold mat over the top for added barrier from seal point; tab also makes it easy to find the opening
  • Absorbs up to 1.41 ounces per bagged absorbent
  • Absorbent-bag combination works on most free aggressive liquids, including acids, bases and unknowns
  • Durable multi-layer construction holds together, even when saturated
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty