Would you like to log in again?
Disposable Tip Pipette Heads
Supplier: Agilent
G5057G G5055G G5057G G5056G G5055G G5056G
77463-582EA 39033 USD InStock
77463-582 CA77463-580 77463-580 CA77463-582 77463-578 CA77463-578
Disposable Tip Pipette Heads
Pipette Tips
Agilent pipette heads are designed to be easily mounted or exchanged and to offer the advantage of being more forgiving than metal-tipped, fixed-tip heads during collisions, increasing uptime during the life of your instrument.
  • Designed to work in two liquid volume ranges: small transfer (ST) and large transfer (LT)
  • Typical %CVs for dispensing 2 µl are 2.1% using DMSO
  • Compatible with most common life science laboratory reagents and, with optimization, CVs better than 5% can be achieved across many liquid types
Disposable-tip pipette heads can aspirate or dispense into one sample well or an array of sample wells (which include a corner and are contiguous). This is primarily used for single-tip cherry picking and occasionally for multiple-tip or array cherry picking.
Agilent disposable-tip pipette heads and tips cover a pipetting volume range from 300 nl to 250 µl with 5% CV. High precision is achieved by optimizing each head for a specific portion of the pipetting volume range.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty