Would you like to log in again?
GemCell™ Plus Xeno-Free Human Serum, World Grade, Gamma Irradiated
Supplier: Gemini Bio-Products
GemCell™ Plus Xeno-Free
100-916G-100
77463-604CS 240 USD InStock
77463-604
GemCell™ Plus Xeno-Free Human Serum, World Grade, Gamma Irradiated
Sera (Serum)
Product has minimum order quantity of 200 units. GemCell™ Plus Xeno-Free, world grade is collected from a maximum of 16 healthy male donors of the AB serotype. Product is defibrinated from source plasma using therapeutic grade recombinant human thrombin. Based on extensive testing and manufacturing process that is Xeno-Free, supports clinical and commercial applications.
  • This product was developed without the use of animal material
  • Custom fill sizes in bottle and bag format offered
  • 0.1 µm sterile-filtered
  • Supports clinical and commerical applications
  • Meet US and EU regulatory requirements
  • Gamma irradiated
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty