Would you like to log in again?
Bravo Disposable Pipette Tips
Supplier: Agilent Technologies
Bravo Lab
11484-312 19133-142 11484-212 10734-212 10734-312 19477-002 19133-212 19134-022 11484-202 19134-002 19134-012 19477-082 11484-322 10734-202 19477-072 10734-302 11484-302 19477-032 19133-102 21740-212 21740-202 19477-022 19133-202 21750-202 19133-112 21750-212 19477-012
77463-618PK 3613 USD InStock
77463-618 77463-640 77463-622 77463-644 77463-642 77463-620 77463-648 77463-704 77463-646 77463-624 77463-608 77463-708 77463-706 77463-652 77463-630 77463-650 77463-710 77463-634 77463-656 77463-612 77463-610 77463-654 77463-632 77463-638 77463-616 77463-614 77463-636
Bravo Disposable Pipette Tips
Pipette Tips
Certified on Bravo and AssayMAP for accuracy and precision.
  • Individual lot certified to meet thorough QC standards
  • Certified free of detectable DNase and RNase activity
  • Certified free of endotoxin (pyrogen) contamination
  • Certified free of adenosine triphosphate (ATP)
  • Certified free of human DNA;Certified free of protease activity
  • Certified free of heavy metals
  • Certified as sterile with a sterility assurance level (SAL) of 10-6
  • Innovative processing and packaging reduce static buildup and improve deck space utilization
Agilent offers disposable pipette tips in both 96- and 384-well formats specially designed, tested, and optimized for the Bravo Automated Liquid Handling Platform and the AssayMAP Bravo heads. All Agilent pipette tips undergo complete QC and functional performance testing for each lot.
The innovative processing and packaging of Bravo robotic tips helps address common concerns about lab disposable pipette tips used in automation, such as static buildup and deck space utilization. Additionally, the Agilent Bravo disposable pipette tips are the only pipette tips that are certified for use on Agilent Bravo Liquid Handling instrumentation, with the assurance that every tip meets our exacting accuracy and precision requirements.
Packaging: Packed as case of 50, except: 21740-202, 21740-212, 21750-202 and 21750-212 which are a case of 90.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty