Would you like to log in again?
Reservoir Microplates, Polypropylene
Supplier: Agilent Technologies
204284-100 201246-100 201256-100 201280-100 204095-100 204093-100 201252-100 201250-100 204484-100 201284-100 204017-100 200686-100 204365-100 204363-100 201254-100 204504-100 201244-100
77463-670PK 194 USD InStock
77463-670 77463-690 77463-684 77463-674 77463-696 77463-694 77463-672 77463-666 77463-688 77463-678 77463-700 77463-686 77463-664 77463-676 77463-698 77463-668 77463-658
Reservoir Microplates, Polypropylene
Microplates HTS Microplates
Polypropylene reservoir microplates for automated liquid handlers and handheld pipettors.
  • Open well format
All microplates are molded using chemical-resistant polymers to ANSI/SLAS standard footprints, with optimized base geometries to minimize dead volumes. They are available with one to 24 cavities per plate, with container heights between 19 and 44 mm, and with clear or black lids. Recommended for method scale-up and scale-down applications, and for creating design of experiment (DoE) arrays.
Packaging: Individually bagged.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty