Would you like to log in again?
Universal Lids for Microplates
Supplier: Agilent Technologies
200856-100 202497-100
77463-680PK 265 USD InStock
77463-680 77463-660
Universal Lids for Microplates
Lids Microplate Lids
Universal lids and cap mats for square microplates.
  • Universal-sized polystyrene lids fit most all microplates
  • Black color is effective for light-sensitive reagents
  • Robot-friendly design
  • Cap mats specifically designed for optimal fit for Agilent square-well microplates
  • Mat material has low permeability to alcohols
Agilent microplate accessories include microplate lids and cap mats. Agilent supplies universal lids for microplates, available in clear or black polystyrene resin. Custom cap mats are available for sealing Agilent 24, 48, and 96-square well microplates. These custom cap mats are molded in Santoprene, a chemical-resistant thermoplastic elastomer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty