Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Storage and Assay Microplates

Supplier: Agilent Technologies

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

204357-100 201240-100 203426-100 204600-100
77463-692PK 172 USD InStock
77463-692 77463-662 77463-682 77463-702
Storage and Assay Microplates
Microplates Deep Well Microplates
For biological assays, low storage temperatures, and filtrate collection applications.

  • Compatible with all major automation platforms and benchtop devices
  • No risk feasibility testing – test a sample in your application
  • Produced to ANSI/SLAS standards, for compatibility with popular labware and devices
  • Molded with chemical resistant polymers, for maintained performance with prolonged exposure to aggressive solutions and solvents
  • Sealable with heat and adhesive seals, or cap mats, for superior performance
  • Low temperature tolerant for refrigerator or freezer storage
  • Autoclavable, can be sterilized at 121 °C, 15 psi for 20 minutes
  • Available in a large range of well volumes, from 58 µl to 10 ml for application scale-up or scale-down
  • Complementary fit to Agilent filter microplates for centrifugation stacking
  • Manufactured in a DNAse/RNase-free ISO 9001 regulated facility

Agilent supplies collection and storage assay microplates for a wide range of reaction chemistries, biological assays, low storage temperatures, and filtrate collection applications. These microplates are molded with medical-grade, chemical-resistant polymers and are available in 24, 48, 96, or 384-well configurations of variable volumes. Agilent also supplies mat seals and lids for these microplates.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty