Would you like to log in again?
PlateLoc Microplate Heat Seals
Supplier: Agilent Technologies
16985-001 17318-001 24214-001 06644-001 24210-001 24212-001
77463-628EA 1539 USD InStock
77463-628 77463-716 77463-606 77463-626 77463-714 77463-712
PlateLoc Microplate Heat Seals
Seals Microplate Seals
Certified for use with the PlateLoc Thermal Microplate Sealer.
  • Choice of aluminum-based or optically clear materials with peelable, pierceable or permanent sealants
  • Seals to match most microplate materials and applications, including polypropylene, polystyrene and COC
Agilent offers a range of PlateLoc microplate heat seals made from aluminum-based or optically clear materials with peelable, pierceable or permanent sealants matching most microplate materials and applications.
Ideal for life science applications that require automated microplate heat sealing, Agilent heat seals keep the PlateLoc Thermal Microplate Sealer up and running reliably to securely heat seal your microplates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty